ENA341295452 Enamine

N-{[3-(aminomethyl)phenyl]methyl}acetamide hydrochloride

  • MDL number MFCD20441634
  • Cas Number : 675112-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-90187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90187
Availability details for
Quantity
  • Availability
  • Add to Cart