ENA341295457 Enamine

4-{[(1H-pyrazol-3-yl)methyl]sulfamoyl}butanoic acid

  • MDL number MFCD14617002
  • Cas Number : 1247819-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92241
Availability details for
Quantity
  • Availability
  • Add to Cart