ENA341295458 Enamine

4-[(4-bromophenoxy)methyl]benzoic acid

  • MDL number MFCD02090847
  • Cas Number : 364623-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92242
Availability details for
Quantity
  • Availability
  • Add to Cart