ENA341295464 Enamine

5-[(2,4,6-trichlorophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD02090892
  • Cas Number : 438220-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92252
Availability details for
Quantity
  • Availability
  • Add to Cart