ENA341295465 Enamine

3-[(diethylamino)methyl]-4-methoxybenzaldehyde

  • MDL number MFCD01822193
  • Cas Number : 128501-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92253
Availability details for
Quantity
  • Availability
  • Add to Cart