ENA341295474 Enamine

3-formylphenyl 1-methyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD02253757
  • Cas Number : 512809-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92262
Availability details for
Quantity
  • Availability
  • Add to Cart