ENA341295475 Enamine

4-(4-ethylphenyl)-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD04967222
  • Cas Number : 1005700-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92263
Availability details for
Quantity
  • Availability
  • Add to Cart