ENA341295479 Enamine

5-[(4-iodophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD03422327
  • Cas Number : 832738-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92267
Availability details for
Quantity
  • Availability
  • Add to Cart