ENA341295487 Enamine

5-(2,4-dichlorophenoxymethyl)furan-2-carbaldehyde

  • MDL number MFCD02253938
  • Cas Number : 438221-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92275
Availability details for
Quantity
  • Availability
  • Add to Cart