ENA341295489 Enamine

5-[(3,5-dimethylphenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD03422322
  • Cas Number : 832740-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92277
Availability details for
Quantity
  • Availability
  • Add to Cart