ENA341295494 Enamine

5-(2-chloro-6-methylphenoxymethyl)furan-2-carbaldehyde

  • MDL number MFCD03422321
  • Cas Number : 832737-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92282

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92282
Availability details for
Quantity
  • Availability
  • Add to Cart