ENA341295496 Enamine

5-[(4-bromo-2-chlorophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD02253763
  • Cas Number : 438219-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92284
Availability details for
Quantity
  • Availability
  • Add to Cart