ENA341295498 Enamine

5-[(4-fluorophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD02253943
  • Cas Number : 438218-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92286
Availability details for
Quantity
  • Availability
  • Add to Cart