ENA341295499 Enamine

3-{[bis(prop-2-en-1-yl)amino]methyl}-4-methoxybenzaldehyde

  • MDL number MFCD04969982
  • Cas Number : 832740-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92287
Availability details for
Quantity
  • Availability
  • Add to Cart