ENA341295506 Enamine

3-[(4-methylphenyl)methoxy]benzaldehyde

  • MDL number MFCD03422459
  • Cas Number : 40359-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92294
Availability details for
Quantity
  • Availability
  • Add to Cart