ENA341295510 Enamine

4-bromo-1-ethyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD03419806
  • Cas Number : 1001519-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92298
Availability details for
Quantity
  • Availability
  • Add to Cart