ENA341295513 Enamine

3,5-dimethyl-1-propyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD07391243
  • Cas Number : 142246-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92301
Availability details for
Quantity
  • Availability
  • Add to Cart