ENA341295517 Enamine

3-[(3-methoxyphenyl)methoxy]benzaldehyde

  • MDL number MFCD03422457
  • Cas Number : 588685-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92305
Availability details for
Quantity
  • Availability
  • Add to Cart