ENA341295521 Enamine

3-[(4-chloro-1H-pyrazol-1-yl)methyl]-4-methoxybenzaldehyde

  • MDL number MFCD01114953
  • Cas Number : 895930-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92309

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92309
Availability details for
Quantity
  • Availability
  • Add to Cart