ENA341295526 Enamine

3-[(3-chlorophenyl)methoxy]benzaldehyde

  • MDL number MFCD03422450
  • Cas Number : 168084-95-5
  • Molecular Weight : 246.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92314
Availability details for
Quantity
  • Availability
  • Add to Cart