ENA341295530 Enamine

5-[(4-ethoxyphenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD03074522
  • Cas Number : 438220-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92318
Availability details for
Quantity
  • Availability
  • Add to Cart