ENA341295534 Enamine

4-[(5-formylfuran-2-yl)methoxy]benzonitrile

  • MDL number MFCD02090905
  • Cas Number : 438221-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92322
Availability details for
Quantity
  • Availability
  • Add to Cart