ENA341295539 Enamine

5-[(2-chloro-4-fluorophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD02253766
  • Cas Number : 438221-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92327
Availability details for
Quantity
  • Availability
  • Add to Cart