ENA341295552 Enamine

3-amino-4-chloro-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06805289
  • Cas Number : 1006451-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92340
Availability details for
Quantity
  • Availability
  • Add to Cart