ENA341295559 Enamine

4-(phenylsulfanyl)benzaldehyde

  • MDL number MFCD01666578
  • Cas Number : 1208-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92347
Availability details for
Quantity
  • Availability
  • Add to Cart