ENA341295564 Enamine

5-[(4-nitro-1H-pyrazol-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06804695
  • Cas Number : 1006457-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92352
Availability details for
Quantity
  • Availability
  • Add to Cart