ENA341295565 Enamine

4-(2,2-difluoroethoxy)-3-nitrobenzoic acid

  • MDL number MFCD12833085
  • Cas Number : 1178286-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92353
Availability details for
Quantity
  • Availability
  • Add to Cart