ENA341295569 Enamine

5-methyl-1-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD03194465
  • Cas Number : 705270-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92357
Availability details for
Quantity
  • Availability
  • Add to Cart