ENA341295578 Enamine

5-[(2,6-dimethylpiperidin-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD15976255
  • Cas Number : 1245808-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92366
Availability details for
Quantity
  • Availability
  • Add to Cart