ENA341295579 Enamine

2-(4-methyl-3-nitro-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD11215411
  • Cas Number : 1174886-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92367
Availability details for
Quantity
  • Availability
  • Add to Cart