ENA341295588 Enamine

1-(butan-2-yl)-4-chloro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06805224
  • Cas Number : 1006451-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92376
Availability details for
Quantity
  • Availability
  • Add to Cart