ENA341295589 Enamine

1-(3,3,3-trifluoropropyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD18262339
  • Cas Number : 1245772-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92377
Availability details for
Quantity
  • Availability
  • Add to Cart