ENA341295603 Enamine

5-[(piperidin-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06803425
  • Cas Number : 893741-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92422
Availability details for
Quantity
  • Availability
  • Add to Cart