ENA341295727 Enamine

2-(difluoromethoxy)-4,5-dimethylbenzaldehyde

  • MDL number MFCD20502019
  • Cas Number : 1354960-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93054
Availability details for
Quantity
  • Availability
  • Add to Cart