ENA341295731 Enamine

4-(4H-1,2,4-triazol-4-yl)butan-1-amine hydrochloride

  • MDL number MFCD20502023
  • Cas Number : 1354950-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93060
Availability details for
Quantity
  • Availability
  • Add to Cart