ENA341295732 Enamine

(4-chloro-2-ethoxyphenyl)methanamine hydrochloride

  • MDL number MFCD20502024
  • Cas Number : 1354950-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...204
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93061

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-93061
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart