ENA341295738 Enamine

1-(3-methyl-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-1-yl)ethan-1-one

  • MDL number MFCD20502035
  • Cas Number : 1354952-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93108
Availability details for
Quantity
  • Availability
  • Add to Cart