ENA341295746 Enamine

tert-butyl 2-methyl-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-1-carboxylate

  • MDL number MFCD20502044
  • Cas Number : 1354954-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93207
Availability details for
Quantity
  • Availability
  • Add to Cart