ENA341295747 Enamine

2-(2,4-dichlorophenyl)-2-(morpholin-4-yl)acetic acid hydrochloride

  • MDL number MFCD20502045
  • Cas Number : 1354949-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93216
Availability details for
Quantity
  • Availability
  • Add to Cart