ENA341295749 Enamine

2,3,4,5-tetrahydro-1$l^{4},5-benzothiazepin-1-one

  • MDL number MFCD20502047
  • Cas Number : 1354961-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93221

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93221
Availability details for
Quantity
  • Availability
  • Add to Cart