ENA341295750 Enamine

2-(3-fluorophenyl)azetidine hydrochloride

  • MDL number MFCD20502048
  • Cas Number : 1354950-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93222
Availability details for
Quantity
  • Availability
  • Add to Cart