ENA341295771 Enamine

5-[(piperidin-1-yl)methyl]thiophene-2-carboxylic acid hydrochloride

  • MDL number MFCD20502070
  • Cas Number : 1354960-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93495
Availability details for
Quantity
  • Availability
  • Add to Cart