ENA341295773 Enamine

2-amino-3,5-difluoro-4-methoxybenzoic acid

  • MDL number MFCD20502072
  • Cas Number : 1354949-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93500

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93500
Availability details for
Quantity
  • Availability
  • Add to Cart