ENA341295779 Enamine

1-(1,1-dioxo-1lambda6-thiolan-3-yl)-2-methyl-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride

  • MDL number MFCD20502078
  • Cas Number : 1354957-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93517
Availability details for
Quantity
  • Availability
  • Add to Cart