ENA341295783 Enamine

3-(aminomethyl)-3-(methylsulfanyl)pentane

  • MDL number MFCD20502082
  • Cas Number : 1354954-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93529
Availability details for
Quantity
  • Availability
  • Add to Cart