ENA341295785 Enamine

3-amino-1lambda4-thiolan-1-one hydrochloride

  • MDL number MFCD20502084
  • Cas Number : 1354957-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93574
Availability details for
Quantity
  • Availability
  • Add to Cart