ENA341295788 Enamine

methyl 2-[4-(chlorosulfonyl)-1H-pyrazol-1-yl]acetate

  • MDL number MFCD20502088
  • Cas Number : 1354952-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93602
Availability details for
Quantity
  • Availability
  • Add to Cart