ENA341295803 Enamine

2-(5-carbamoyl-1H-pyrrole-3-sulfonamido)acetic acid

  • MDL number MFCD20502106
  • Cas Number : 1354961-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93777
Availability details for
Quantity
  • Availability
  • Add to Cart