ENA341295806 Enamine

5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazin-3-ylmethanol dihydrochloride

  • MDL number MFCD20502110
  • Cas Number : 1354953-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93864

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-93864
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart