ENA341295813 Enamine

methyl 2-amino-3-fluoro-4-methoxybenzoate

  • MDL number MFCD20502117
  • Cas Number : 1180497-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93900

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-93900
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart